Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The Human Siglec-9 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Siglec-9.
Specifications
Specifications
| Quantity | 1 Kit (for 15 Plates) |
| Species | Human |
| Conjugate | Biotin |
| Assay Range | 62.50 to 4000 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 27180 |
| Synonym | CD329, CDw329, FOAP-9, OBBP-Like, Protein FOAP-9, sialic acid binding Ig-like lectin 9, sialic acid-binding Ig-like lectin 9, siglec-9, Siglec9 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
