Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Sigma-1 R Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1076100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1076100 100 μg
MAB1076SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1076100 Supplier R&D Systems Supplier No. MAB1076100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Sigma-1 R/OPRS1 Monoclonal specifically detects Sigma-1 R/OPRS1 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen Sigma-1 R/OPRS1
Applications Immunocytochemistry
Classification Monoclonal
Clone 1319A
Conjugate Unconjugated
Dilution Immunocytochemistry 3-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q99720
Gene Alias Aging-associated gene 8 protein, ALS16, FLJ25585, hSigmaR1, OPRS1, SIG-1R, sigma 1, Sigma 1-type opioid receptor, sigma non-opioid intracellular receptor 1, sigma1 receptor, sigma1R, Sigma-1R, sigma1-receptor, SIGMAR1, SR31747 binding protein 1, SR31747-binding protein, SRBP, SR-BP, SR-BP1, SRBPMGC3851, type I sigma receptor
Host Species Rabbit
Immunogen Synthetic peptide containing human Sigma-1 R, Accession # Q99720
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 10280
Test Specificity Detects human Sigma-1 R in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.