Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SIRP alpha/CD172a Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB4546S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4546S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB4546S100 Supplier R&D Systems Supplier No. FAB4546S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SIRP alpha/CD172a Monoclonal specifically detects SIRP alpha/CD172a in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen SIRP alpha/CD172a
Applications Flow Cytometry
Classification Monoclonal
Clone 602411
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P78324
Gene Alias Bit, BITbrain-immunoglobulin-like molecule with tyrosine-based activation motifs, Brain Ig-like molecule with tyrosine-based activation motifs, CD172 antigen-like family member A, CD172a, CD172a antigen, Inhibitory receptor SHPS-1, Macrophage fusion receptor, MFR, MFRtyrosine phosphatase SHP substrate 1, MYD1, MYD-1, MyD-1 antigen, P84, protein tyrosine phosphatase, non-receptor type substrate 1, PTPNS1, SHP substrate 1, SHPS1, SHPS-1, SHPS1CD172A, signal-regulatory protein alpha, Signal-regulatory protein alpha-1, Signal-regulatory protein alpha-2, Signal-regulatory protein alpha-3, SIRPA, SIRPalpha, SIRP-ALPHA-1, Sirp-alpha-2, SIRPalpha2, Sirp-alpha-3, SIRPtyrosine-protein phosphatase non-receptor type substrate 1
Gene Symbols SIRPA
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human SIRP alpha/CD172a Gly27-Asn370 (predicted) Accession # P78324
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140885
Test Specificity Detects human SIRP alpha/CD172a in direct ELISAs and Western blots. In direct ELISAs, 50-100% cross-reactivity with recombinant human (rh) SIRP beta 1 and no cross-reactivity with rhSIRP beta 2 is observed. In Western blots, approximately 10% cross-reactivity with rhSIRP beta 1 and no cross-reactivity with rhSIRP beta 2 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.