Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SIRP beta 1/CD172b Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB20961100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB20961100 100 μg
MAB20961SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB20961100 Supplier R&D Systems Supplier No. MAB20961100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SIRP beta 1/CD172b Monoclonal specifically detects SIRP beta 1/CD172b in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen SIRP beta 1/CD172b
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 308908
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. O00241.4
Gene Alias CD172 antigen-like family member B, CD172b, CD172b antigen, DKFZp686A05192, FLJ26614, signal-regulatory protein beta 1, signal-regulatory protein beta-1, SIRPB1, SIRP-BETA-1, SIRP-beta-1 isoform 3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human SIRP beta 1/CD172b, Gly26-Ala369 (Arg53His and Ala363Pro), Accession # O00241.4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10326
Test Specificity Detects human SIRP beta 1/CD172b in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.