Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SIRP beta 1/gamma Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2096U100UG
This item is not returnable.
View return policy
Description
SIRP beta 1/gamma Monoclonal specifically detects SIRP beta 1/gamma in Human samples. It is validated for Flow Cytometry.Specifications
SIRP beta 1/gamma | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
O00241.4 | |
SIRPB1 | |
Mouse myeloma cell line NS0-derived recombinant human SIRP beta 1/CD172b Gly26-Ala369 (Arg53His and Ala363Pro) Accession # O00241.4 | |
RUO | |
10326 | |
Human | |
IgG2a |
Flow Cytometry | |
308906 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD172 antigen-like family member B, CD172b, CD172b antigen, DKFZp686A05192, FLJ26614, signal-regulatory protein beta 1, signal-regulatory protein beta-1, SIRPB1, SIRP-BETA-1, SIRP-beta-1 isoform 3 | |
Mouse | |
100 μg | |
Primary | |
Detects human SIRP beta 1/CD172b in direct ELISAs and Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction