Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SIRP beta 1/gamma Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2096R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2096R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2096R100 Supplier R&D Systems Supplier No. FAB2096R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SIRP beta 1/gamma Monoclonal specifically detects SIRP beta 1/gamma in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen SIRP beta 1/gamma
Applications Flow Cytometry
Classification Monoclonal
Clone 308906
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O00241.4
Gene Alias CD172 antigen-like family member B, CD172b, CD172b antigen, DKFZp686A05192, FLJ26614, signal-regulatory protein beta 1, signal-regulatory protein beta-1, SIRPB1, SIRP-BETA-1, SIRP-beta-1 isoform 3
Gene Symbols SIRPB1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human SIRP beta 1/CD172b Gly26-Ala369 (Arg53His and Ala363Pro) Accession # O00241.4
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10326
Test Specificity Detects human SIRP beta 1/CD172b in direct ELISAs and Western blots.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.