Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SIRP beta 1/gamma Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | SIRP beta 1/gamma |
---|---|
Clone | 308906 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Description
SIRP beta 1/gamma Monoclonal specifically detects SIRP beta 1/gamma in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
SIRP beta 1/gamma | |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
O00241.4 | |
10326 | |
Mouse myeloma cell line NS0-derived recombinant human SIRP beta 1, Gly26-Ala369 (Arg53His and Ala363Pro), Accession # O00241.4 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human SIRP beta 1 and human SIRP gamma in direct ELISAs and Western blots. |
308906 | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD172 antigen-like family member B, CD172b, CD172b antigen, DKFZp686A05192, FLJ26614, signal-regulatory protein beta 1, signal-regulatory protein beta-1, SIRPB1, SIRP-BETA-1, SIRP-beta-1 isoform 3 | |
SIRPB1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title