Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SIRP beta 2 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  R&D Systems FAB99981T100UG

Catalog No. FAB99981T10


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

SIRPB2 Monoclonal specifically detects SIRPB2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

SIRPB2
Monoclonal
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
dJ776F14.2, Protein Tyrosine Phosphatase Non-Receptor Type Substrate 1-Like 3, Protein Tyrosine Phosphatase Non-Receptor Type Substrate Protein, Protein Tyrosine Phosphatase, Non-Receptor Type 1-Like, Protein Tyrosine Phosphatase, Non-Receptor Type Substrate 1-Like 3, PTPN1L, PTPNS1L3, Signal-Regulatory Protein Beta 2, Signal-Regulatory Protein Beta-2, SIRP-beta-2, SIRPG
HEK293 human embryonic kidney cell line, HEK293 derived Human SIRP beta 2, Gln33-Gly287, Accession # Q5JXA9
100 μg
284759
Human
Supernatant
Flow Cytometry
1008702
Flow Cytometry 0.25-1 μg/106 cells
Q5JXA9
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human SIRP beta 2 in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.