Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Sirtuin 1/SIRT1 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC7714S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC7714S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC7714S100 Supplier R&D Systems Supplier No. IC7714S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sirtuin 1/SIRT1 Monoclonal specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Sirtuin 1/SIRT1
Applications Flow Cytometry
Classification Monoclonal
Clone 834918
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q96EB6
Gene Alias EC 3.5.1, S. cerevisiae, homolog) 1, SIR2alpha, SIR2L1, SIR2-like protein 1, SIRT1, sirtuin 1
Host Species Mouse
Immunogen E. coli-derived recombinant human Sirtuin 1/SIRT1 Met1-Ser747 Accession # Q96EB6
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23411
Test Specificity Detects human Sirtuin 1/SIRT1 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Sirtuin 2, 3, 5, or 6 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.