Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Sirtuin 1/SIRT1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7714
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7714 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7714 Supplier R&D Systems Supplier No. MAB7714
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sirtuin 1/SIRT1 Monoclonal specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen Sirtuin 1/SIRT1
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 834918
Concentration LYOPH
Conjugate Unconjugated
Dilution Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q96EB6
Gene Alias EC 3.5.1, S. cerevisiae, homolog) 1, SIR2alpha, SIR2L1, SIR2-like protein 1, SIRT1, sirtuin 1
Gene Symbols SIRT1
Host Species Mouse
Immunogen E. coli-derived recombinant human Sirtuin 1/SIRT1 Met1-Ser747 Accession # Q96EB6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23411
Test Specificity Detects human Sirtuin 1/SIRT1 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Sirtuin 2, 3, 5, or 6 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.