Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Sirtuin 1/SIRT1 ELISA Kit (Colorimetric)
SDP

Catalog No. NB013371 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013371 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013371 Supplier Novus Biologicals™ Supplier No. NBP280300
Add to Cart
Add to Cart

This kit recognizes Human SIRT1 in samples.

No significant cross-reactivity or interference between Human SIRT1 and analogues was observed.

Specifications

Accession Number Q96EB6, Q96EB6
Assay Range 0.313 to 20 ng/mL
Assay Sensitivity 0.188 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 23411
Gene Symbol SIRT1
Host Species Human
Quantity 1 Kit
Research Discipline Apoptosis, Chromatin Research, Cognition and Behavior, DNA Repair, Epigenetics, Phospho Specific
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target Sirtuin 1/SIRT1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.