Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Sirtuin 2/SIRT2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4358SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4358SP 25 μg
MAB4358 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4358SP Supplier R&D Systems Supplier No. MAB4358SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sirtuin 2/SIRT2 Monoclonal specifically detects Sirtuin 2/SIRT2 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Sirtuin 2/SIRT2
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 506611
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8IXJ6
Gene Alias EC 3.5.1, EC 3.5.1.-, FLJ35621, FLJ37491, S.cerevisiae, homolog) 2, SIR2L2, SIRT2, sirtuin 2, sirtuin type 2
Gene Symbols SIRT2
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Sirtuin 2/SIRT2 Ala2-Gln389 Accession # Q8IXJ6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22933
Test Specificity Detects human Sirtuin 2/SIRT2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human SIRT1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.