Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Sirtuin 2/SIRT2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4358SP
This item is not returnable.
View return policy
Description
Sirtuin 2/SIRT2 Polyclonal specifically detects Sirtuin 2/SIRT2 in Human samples. It is validated for Western Blot.Specifications
| Sirtuin 2/SIRT2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.5.1, EC 3.5.1.-, FLJ35621, FLJ37491, S.cerevisiae, homolog) 2, SIR2L2, SIRT2, sirtuin 2, sirtuin type 2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 22933 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q8IXJ6 | |
| SIRT2 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Sirtuin 2/SIRT2 Ala2-Gln389 (predicted) Accession # Q8IXJ6 | |
| 25 μg | |
| Primary | |
| Detects human Sirtuin 2/SIRT2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human SIRT1 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction