Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SKAP55 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4264SP
This item is not returnable.
View return policy
Description
SKAP55 Polyclonal specifically detects SKAP55 in Human samples. It is validated for Western Blot.Specifications
SKAP55 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
SCAP1, SCAP1pp55, SKAP1, SKAP55SKAP-55, src family associated phosphoprotein 1, Src family-associated phosphoprotein 1, src kinase associated phosphoprotein 1, src kinase-associated phosphoprotein 1, Src kinase-associated phosphoprotein of 55 kDa | |
Goat | |
Affinity Purified | |
RUO | |
8631 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
NP_001068567 | |
SKAP1 | |
E. coli-derived recombinant human SKAP55 Met1-Arg358 Accession # NP_001068567 | |
25 μg | |
Primary | |
Detects human SKAP55 in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction