Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SLAM/CD150 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $351.50
Specifications
Antigen | SLAM/CD150 |
---|---|
Clone | 123303 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB16421SP
![]() |
R&D Systems
MAB16421SP |
25 μg |
Each for $200.50
|
|
|||||
MAB16421100
![]() |
R&D Systems
MAB16421100 |
100 μg |
Each for $351.50
|
|
|||||
Description
SLAM/CD150 Monoclonal specifically detects SLAM/CD150 in Human samples. It is validated for ELISA.Specifications
SLAM/CD150 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
CD 150, CD150, CD150IPO-3, CDw150, IPO-3, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen, SLAMF1 | |
Mouse myeloma cell line NS0-derived recombinant human SLAM/CD150, Ala21-Pro237, Accession # Q13291 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human SLAM/CD150 in direct ELISAs. |
123303 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
Q13291 | |
6504 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title