Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SLC14A1 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8238R100UG
This item is not returnable.
View return policy
Description
SLC14A1 Monoclonal specifically detects SLC14A1 in Human samples. It is validated for Flow Cytometry.Specifications
SLC14A1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q13336 | |
SLC14A1 | |
NS0 mouse myeloma cell line transfected with human SLC14A1 Accession # Q13336 | |
RUO | |
6563 | |
Human | |
IgG2b |
Flow Cytometry | |
888418 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
FLJ33745, HsT1341, HUT11, JK, JKFLJ41687, RACH1, RACH2, solute carrier family 14 (urea transporter), member 1 (Kidd blood group), Solute carrier family 14 member 1, truncated urea transporter, urea transporter 1, urea transporter JK glycoprotein, Urea transporter, erythrocyte, urea transporter-B1, UT1, UT1UT-B1, UT-B1, UTE, UTEblood group Kidd urea transporter | |
Mouse | |
100 μg | |
Primary | |
Detects human SLC14A1 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction