Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SLC22A1 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6469N100UG
This item is not returnable.
View return policy
Description
SLC22A1 Monoclonal antibody specifically detects SLC22A1 in Human samples. It is validated for Flow CytometrySpecifications
SLC22A1 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
hOCT1, OCT1, oct1_cds, OCT1solute carrier family 22 member 1, Organic cation transporter 1, solute carrier family 22 (organic cation transporter), member 1 | |
NS0 mouse myeloma cell line transfected with human SLC22A1, Met1-Thr554, Accession # NP_003048 | |
100 μg | |
Primary | |
Reconstitution default not known | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Flow Cytometry | |
609222 | |
Flow Cytometry 0.25-1 ug/106 cells | |
NP_003048 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6580 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction