Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SLC22A1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6469SP
This item is not returnable.
View return policy
Description
SLC22A1 Monoclonal specifically detects SLC22A1 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
SLC22A1 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells | |
NP_003048 | |
SLC22A1 | |
NS0 mouse myeloma cell line transfected with human SLC22A1 Met1-Thr554 Accession # NP_003048 | |
25 μg | |
Primary | |
Detects human SLC22A1 in Western blots. | |
Human | |
Purified |
Western Blot, Flow Cytometry | |
609222 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
hOCT1, OCT1, oct1_cds, OCT1solute carrier family 22 member 1, Organic cation transporter 1, solute carrier family 22 (organic cation transporter), member 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6580 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction