Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC25A15 Full-length ORF (NP_055067.1, 1 a.a. - 301 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7098514
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene is a member of the mitochondrial carrier family. The encoded protein transports ornithine across the inner mitochondrial membrane from the cytosol to the mitochondrial matrix. The protein is an essential component of the urea cycle, and functions in ammonium detoxification and biosynthesis of the amino acid arginine. Mutations in this gene result in hyperornithinemia-hyperammonemia-homocitrullinuria (HHH) syndrome. There is a pseudogene of this locus on the Y chromosome

Sequence: MKSNPAIQAAIDLTAGAAGGTACVLTGQPFDTMKVKMQTFPDLYRGLTDCCLKTYSQVGFRGFYKGTSPALIANIAENSVLFMCYGFCQQVVRKVAGLDKQAKLSDLQNAAAGSFASAFAALVLCPTELVKCRLQTMYEMETSGKIAKSQNTVWSVIKSILRKDGPLGFYHGLSSTLLREVPGYFFFFGGYELSRSFFASGRSKDELGPVPLMLSGGVGGICLWLAVYPVDCIKSRIQVLSMSGKQAGFIRTFINVVKNEGITALYSGLKPTMIRAFPANGALFLAYEYSRKLMMNQLEAY

Specifications

Accession Number NP_055067.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10166
Molecular Weight (g/mol) 59.1kDa
Name SLC25A15 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MKSNPAIQAAIDLTAGAAGGTACVLTGQPFDTMKVKMQTFPDLYRGLTDCCLKTYSQVGFRGFYKGTSPALIANIAENSVLFMCYGFCQQVVRKVAGLDKQAKLSDLQNAAAGSFASAFAALVLCPTELVKCRLQTMYEMETSGKIAKSQNTVWSVIKSILRKDGPLGFYHGLSSTLLREVPGYFFFFGGYELSRSFFASGRSKDELGPVPLMLSGGVGGICLWLAVYPVDCIKSRIQVLSMSGKQAGFIRTFINVVKNEGITALYSGLKPTMIRAFPANGALFLAYEYSRKLMMNQLEAY
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias D13S327/HHH/ORC1/ORNT1
Common Name SLC25A15
Gene Symbol SLC25A15
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.