Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SLC38A4 Partial ORF (NP_060488, 245 a.a. - 331 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7100922
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

SLC38A4 is found predominantly in liver and transports both cationic and neutral amino acids. The transport of cationic amino acids by SLC38A4 is Na(+) and pH independent, while the transport of neutral amino acids is Na(+) and pH dependent (Hatanaka et al., 2001 [PubMed 11342143]).[supplied by OMIM]

Sequence: FQIPCPLPVLDHSVGNLSFNNTLPMHVVMLPNNSESSDVNFMMDYTHRNPAGLDENQAKGSLHDSGVEYEAHSDDKCEPKYFVFNSR

Specifications

Accession Number NP_060488
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55089
Molecular Weight (g/mol) 35.31kDa
Name SLC38A4 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen FQIPCPLPVLDHSVGNLSFNNTLPMHVVMLPNNSESSDVNFMMDYTHRNPAGLDENQAKGSLHDSGVEYEAHSDDKCEPKYFVFNSR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ATA3/FLJ10191/MGC126876/NAT3/PAAT
Common Name SLC38A4
Gene Symbol SLC38A4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.