Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SLC5A5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8367
This item is not returnable.
View return policy
Description
SLC5A5/Sodium Iodide Symporter Monoclonal specifically detects SLC5A5/Sodium Iodide Symporter in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
SLC5A5/Sodium Iodide Symporter | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NIS, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6528 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Flow Cytometry, CyTOF | |
902320 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Q92911 | |
SLC5A5 | |
NS0 mouse myeloma cell line transfected with human SLC5A5 Accession # Q92911 | |
100 μg | |
Primary | |
Stains human SLC5A5 transfectants but not irrelevant transfectants in flow cytometry. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction