Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SLC6A4/5-HTTLPR Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB17292V100UG
This item is not returnable.
View return policy
Description
SLC6A4/5-HTTLPR/Serotonin transporter Monoclonal antibody specifically detects SLC6A4/5-HTTLPR/Serotonin transporter in Human samples. It is validated for Flow CytometrySpecifications
SLC6A4/5-HTTLPR/Serotonin transporter | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
5HT transporter, 5-HT Transporter, 5-HTT, 5HTTLPR, 5-HTTLPR, hSERT, HTT5-hydroxytryptamine transporter, Na+/Cl- dependent serotonin transporter, OCD1, Serotonin Transporter 1, SERT, sodium-dependent serotonin transporter, solute carrier family 6 (neurotransmitter transporter, serotonin), member 4,5-HTTLPR, Solute carrier family 6 member 4,5HTT | |
Human SLC6A4/5-HTTLPR synthetic peptide, Accession # P31645 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
632705 | |
Flow Cytometry | |
P31645 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6532 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction