Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SLC7A5/LAT1 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Catalog No. FAB10390G10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10390G10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10390G10 Supplier R&D Systems Supplier No. FAB10390G100UG

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SLC7A5/LAT1 Monoclonal specifically detects SLC7A5/LAT1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen SLC7A5/LAT1
Applications Flow Cytometry
Classification Monoclonal
Clone BU53
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q01650
Gene Alias 4F2 LC, 4F2LC, CD98, CD98LC, D16S469EL-type amino acid transporter 1, E16, Integral membrane protein E16, large neutral amino acids transporter 1, large neutral amino acids transporter small subunit 1, LAT1, LAT1hLAT1, MPE16, MPE16CD98 light chain, sodium-independent neutral amino acid transporter LAT1,4F2LC, solute carrier family 7 (cationic amino acid transporter, y+ system), member 5,4F2 light chain, Solute carrier family 7 member 5, TA1, y+ system cationic amino acid transporter
Host Species Mouse
Immunogen Plasma cell line 8226, Accession # Q01650
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8140
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.