Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Slit2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB54443100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB54443100 100 μg
MAB54443SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB54443100 Supplier R&D Systems Supplier No. MAB54443100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Slit2 Monoclonal specifically detects Slit2 in Human samples. It is validated for ELISA.

Specifications

Antigen Slit2
Applications ELISA
Classification Monoclonal
Clone 984724
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. O94813-1
Gene Alias FLJ14420, SLIL3, slit (Drosophila) homolog 2, slit homolog 2 (Drosophila), slit homolog 2 protein, Slit-2SLIL3
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human Slit2, Thr1122-Ser1529, Accession No. O94813-1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 9353.0
Test Specificity Detects human Slit2 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.