Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SLITRK1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3009SP
This item is not returnable.
View return policy
Description
SLITRK1 Polyclonal specifically detects SLITRK1 in Human samples. It is validated for Western Blot.Specifications
SLITRK1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ54428, KIAA1910KIAA0918, leucine rich repeat containing 12, Leucine-rich repeat-containing protein 12, LRRC12, RP11-395N17.1, SLIT and NTRK-like family, member 1, SLIT and NTRK-like protein 1, slit and trk like gene 1, TTM | |
Goat | |
Affinity Purified | |
RUO | |
114798 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Q96PX8 | |
SLITRK1 | |
Mouse myeloma cell line NS0-derived recombinant human SLITRK1 (R&D Systems, Catalog # 3009-SK) Ala16-Ser616 Accession # Q96PX8 | |
25 μg | |
Primary | |
Detects human SLITRK1 in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with recombinant human (rh) SLITRK2, rhSLITRK4, and rhSLITRK5 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction