Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SLITRK1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB30091
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB30091 100 μg
MAB30091SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB30091 Supplier R&D Systems Supplier No. MAB30091
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SLITRK1 Monoclonal specifically detects SLITRK1 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen SLITRK1
Applications Immunohistochemistry
Classification Monoclonal
Clone 327829
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q96PX8
Gene Alias FLJ54428, KIAA1910KIAA0918, leucine rich repeat containing 12, Leucine-rich repeat-containing protein 12, LRRC12, RP11-395N17.1, SLIT and NTRK-like family, member 1, SLIT and NTRK-like protein 1, slit and trk like gene 1, TTM
Gene Symbols SLITRK1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human SLITRK1 Ala16-Ser616 Accession # Q96PX8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114798
Test Specificity Detects human SLITRK1 in direct ELISAs. In direct ELISAs, no cross-reactivity with reactivity with SLITRK2, 3, 4, or 5 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.