Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SLITRK2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2657
This item is not returnable.
View return policy
Description
SLITRK2 Polyclonal specifically detects SLITRK2 in Human samples. It is validated for Western Blot.Specifications
| SLITRK2 | |
| Polyclonal | |
| Unconjugated | |
| Q9H156 | |
| SLITRK2 | |
| Mouse myeloma cell line NS0-derived recombinant human SLITRK2 Thr19-Pro617 Accession # Q9H156 | |
| 100 μg | |
| Primary | |
| Detects human SLITRK2 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) SLITRK4 is observed and less than 2% cross-reactivity with rhSLITRK1 and rhSLITRK5 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CXorf2, SLITL1, SLITRK2 SLIT and NTRK-like family, member 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 84631 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction