Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SLITRK5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2587
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2587 100 μg
AF2587SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2587 Supplier R&D Systems Supplier No. AF2587
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

SLITRK5 Polyclonal specifically detects SLITRK5 in Human samples. It is validated for Western Blot.

Specifications

Antigen SLITRK5
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. O94991
Gene Alias bA364G4.2, FLJ58374, KIAA0918, leucine-rich repeat-containing protein 11, LRRC11, SLIT and NTRK-like family, member 5, SLIT and NTRK-like protein 5
Gene Symbols SLITRK5
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human SLITRK5 Glu39-Val665 Accession # O94991
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26050
Test Specificity Detects human SLITRK5 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) SLITRK1, rhSLITRK2, rhSLITRK3, rhSLITRK4 and rhSLITRK6 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.