Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human SLPI DuoSet ELISA
SDP

Catalog No. DY127405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY127405 1 Kit (for 5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY127405 Supplier R&D Systems™ Supplier No. DY127405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols

Human SLPI DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human SLPI DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human SLPI.

Specifications

Description ALK1, ALPProtease inhibitor WAP4, antileukoproteinase, BLPISeminal proteinase inhibitor, HUSI, HUSI-I, Mucus proteinase inhibitor, secretory leukocyte peptidase inhibitor, Secretory leukocyte protease inhibitor, secretory leukocyte protease inhibitor (antileukoproteinase), WAP four-disulfide core domain protein 4, WAP4HUSI-1, WFDC4MPI
Product Type ELISA Kit
Quantity 1 Kit (for 5 Plates)
Species Human
Cross Reactivity Not observed
Format 96-well Strip Plate
For Use With (Application) ELISA
Gene ID (Entrez) 6590
Sample Type Cell Culture Supernatant, Serum, Plasma
Kit Contents Capture antibody, Detection antibody, Recombinant standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.