Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SLURP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | SLURP1 |
---|---|
Clone | 569317 |
Concentration | LYOPH |
Dilution | Immunohistochemistry 8-25 ug/mL |
Applications | Immunohistochemistry |
Description
SLURP1 Monoclonal specifically detects SLURP1 in Human samples. It is validated for Immunohistochemistry.Specifications
SLURP1 | |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
P55000 | |
57152 | |
E. coli-derived recombinant human SLURP1 Ala22-Leu103 Accession # P55000 | |
Primary | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Detects human SLURP1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse SLURP1 is observed. |
569317 | |
Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ANUPARS(component B)-81/S, ARS component B, ArsB, ARSlymphocyte antigen 6-like secreted, LY6LS, MDMsecreted Ly-6/uPAR-related protein 1, neoplastic urinary protein, secreted LY6/PLAUR domain containing 1, secreted Ly6/uPAR related protein 1, SLURP-1 | |
SLURP1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title