Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SMAC/Diablo Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF7891SP
This item is not returnable.
View return policy
Description
SMAC/Diablo Polyclonal specifically detects SMAC/Diablo in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SMAC/Diablo | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
0610041G12Rik, Diablo, diablo, IAP-binding mitochondrial protein, Direct IAP-binding protein with low pI, mitochondrial Smac protein, Second mitochondria-derived activator of caspase, Smac, SMACmitochondrial | |
Goat | |
Affinity Purified | |
RUO | |
56616 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Q9NR28 | |
DIABLO | |
E. coli-derived recombinant human SMAC/Diablo Ala56-Asp239 Accession # Q9NR28 | |
25 μg | |
Primary | |
Detects human, mouse, and rat SMAC/Diablo in direct ELISAs and Western blots. No cross-reactivity with recombinant human NME3 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction