Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SMAC/Diablo Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF789
This item is not returnable.
View return policy
Description
SMAC/Diablo Polyclonal specifically detects SMAC/Diablo in Human samples. It is validated for Western Blot.Specifications
SMAC/Diablo | |
Polyclonal | |
Biotin | |
Q9NR28 | |
DIABLO | |
E. coli-derived recombinant human SMAC Ala56-Asp239 Accession # Q9NR28 | |
50 μg | |
Primary | |
Detects human SMAC in Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
0610041G12Rik, Diablo, diablo, IAP-binding mitochondrial protein, Direct IAP-binding protein with low pI, mitochondrial Smac protein, Second mitochondria-derived activator of caspase, Smac, SMACmitochondrial | |
Goat | |
Affinity Purified | |
RUO | |
56616 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction