Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Smad8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2309
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2309 100 μg
AF2309SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2309 Supplier R&D Systems Supplier No. AF2309
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Smad8 Polyclonal specifically detects Smad8 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Smad8
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. NP_005896
Gene Symbols SMAD9
Host Species Goat
Immunogen E. coli-derived recombinant human Smad8 Ala158-Ser233 Accession # NP_005896
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4093
Test Specificity Detects human Smad8 in direct ELISAs and Western blots. In direct ELISAs, approximately 90% cross-reactivity with recombinant (rh) Smad1 is observed, approximately 20% cross-reactivity with rhSmad4 is observed, and less than 1% cross-reactivity with rhSmad5, rhSmad7, and rhDNAK is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.