Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SMAGP Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3959
This item is not returnable.
View return policy
Description
SMAGP Polyclonal specifically detects SMAGP in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SMAGP | |
Polyclonal | |
Unconjugated | |
Q0VAQ4 | |
SMAGP | |
E. coli-derived recombinant human SMAGP Thr2-Thr34, Phe56-Ile97 Accession # Q0VAQ4 | |
100 μg | |
Primary | |
Detects human SMAGP in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant rat SMAGP is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
hSMAGP, LOC57228, MGC149453, MGC149454, small cell adhesion glycoprotein, small cell transmembrane and glycosylated protein, small trans-membrane and glycosylated protein, Small transmembrane and glycosylated protein | |
Goat | |
Affinity Purified | |
RUO | |
57228 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction