Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SMARCA5/SNF2H Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF6825SP
This item is not returnable.
View return policy
Description
SMARCA5/SNF2H Polyclonal specifically detects SMARCA5/SNF2H in Human samples. It is validated for Immunohistochemistry.Specifications
SMARCA5/SNF2H | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.6.1, EC 3.6.4.-, hISWI, hSNF2HSWI/SNF-related matrix-associated actin-dependent regulator of chromatin A5, ISWI, SNF2H, SNF2Hsucrose nonfermenting-like 5, subfamily a, member 5, Sucrose nonfermenting protein 2 homolog, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 5, WCRF135 | |
Goat | |
Affinity Purified | |
RUO | |
8467 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Immunohistochemistry | |
LYOPH | |
Immunohistochemistry 5-15 ug/mL | |
O60264 | |
SMARCA5 | |
E. coli-derived recombinant human SAMRCA5/SNF2H Asp77-Val185 Accession # O60264 | |
25 μg | |
Primary | |
Detects human SMARCA5/SNF2H in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) SMARCA1 and rhSMARC4 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction