Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SMOC-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5140
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5140 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5140 Supplier R&D Systems Supplier No. MAB5140
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SMOC-2 Monoclonal specifically detects SMOC-2 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen SMOC-2
Applications Immunohistochemistry
Classification Monoclonal
Clone 667713
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9H3U7
Gene Alias bA270C4A.1, bA37D8.1, dJ421D16.1, MST117, MSTP117, MSTP140, SMAP2, SMAP-2, SMAP2Secreted modular calcium-binding protein 2, SMOC2, SMOC-2, smooth muscle associated protein 2, Smooth muscle-associated protein 2, SPARC related modular calcium binding 2, SPARC-related modular calcium-binding protein 2, thyroglobulin type-1 repeat containing protein
Gene Symbols SMOC2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human SMOC-2 Gln22-Gly446 Accession # Q9H3U7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64094
Test Specificity Detects human SMOC-2 in direct ELISAs and Western blots. In Western blots, approximately 15% cross-reactivity with recombinant human (rh) SMOC-1 and no cross-reactivity with recombinant mouse SMOC-2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.