Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Smoothelin B Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | Smoothelin |
---|---|
Clone | 885424 |
Concentration | LYOPH |
Dilution | Immunohistochemistry 5-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Immunohistochemistry, Flow Cytometry, CyTOF |
Description
Smoothelin Monoclonal specifically detects Smoothelin in Human samples. It is validated for Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
Smoothelin | |
LYOPH | |
Immunohistochemistry, Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
P53814 | |
6525 | |
E. coli-derived recombinant human Smoothelin B Met1-Leu132 Accession # P53814 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Smoothelin B in direct ELISAs. |
885424 | |
Immunohistochemistry 5-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ35365, FLJ38597, smoothelin, SMSMO, SMTN | |
SMTN | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title