Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SMURF1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$188.28 - $520.35

Specifications

Antigen SMURF1
Clone 530131
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB9507SP
GREENER_CHOICE
View Documents
R&D Systems
MAB9507SP
25 μg
Each for $188.28
Only null left
Add to Cart
 
MAB9507100
GREENER_CHOICE
View Documents
R&D Systems
MAB9507100
100 μg
Each for $520.35
Only null left
Add to Cart
 
Description

Description

SMURF1 Monoclonal specifically detects SMURF1 in Human samples. It is validated for Western Blot, Simple Western.
Specifications

Specifications

SMURF1
Western Blot 1 ug/mL, Simple Western 10 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
E3 ubiquitin ligase SMURF1, EC 6.3.2, EC 6.3.2.-, hSMURF1, KIAA1625, KIAA1625E3 ubiquitin-protein ligase SMURF1, SMAD specific E3 ubiquitin protein ligase 1, SMAD ubiquitination regulatory factor 1, Smad-specific E3 ubiquitin ligase 1, SMAD-specific E3 ubiquitin-protein ligase 1
E. coli-derived recombinant human SMURF1 Met496-Glu757 Accession # Q9HCE7
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human SMURF1 in direct ELISAs.
530131
Western Blot
Unconjugated
Mouse
Human
Q9HCE7
57154
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only (RUO)

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.