Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Snail Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Supplier: R&D Systems AF3639
This item is not returnable.
View return policy
Description
Snail Polyclonal specifically detects Snail in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.Specifications
Snail | |
Polyclonal | |
Unconjugated | |
O95863 | |
SNAI1 | |
E. coli-derived recombinant human Snail Pro2-Arg264 Accession # O95863 | |
100 μg | |
Primary | |
Detects human Snail in direct ELISAs and Western blots. | |
Human | |
IgG |
ChIP Assay, Flow Cytometry, Immunocytochemistry | |
LYOPH | |
Western Blot 0.5 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL | |
Protein sna, Protein snail homolog 1, SLUGH2, SNA, SNAH, SNAHdJ710H13.1, SNAI1, snail 1 (drosophila homolog), zinc finger protein, snail 1 homolog, snail 1 zinc finger protein, snail 1, zinc finger protein, snail homolog 1 (Drosophila), zinc finger protein SNAI1 | |
Goat | |
Affinity Purified | |
RUO | |
6615 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction