Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SNAP23 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6306SP
This item is not returnable.
View return policy
Description
SNAP23 Polyclonal specifically detects SNAP23 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
SNAP23 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HsT17016, SNAP-23, SNAP23A, SNAP23B, synaptosomal-associated protein 23, synaptosomal-associated protein, 23kD, synaptosomal-associated protein, 23kDa, Vesicle-membrane fusion protein SNAP-23 | |
Sheep | |
Affinity Purified | |
RUO | |
8773 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
O00161 | |
SNAP23 | |
E. coli-derived recombinant human SNAP23 Asp146-Ser211 Accession # O00161 | |
25 μg | |
Primary | |
Detects human SNAP23 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse SNAP23 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction