Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SNAP23 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6306
This item is not returnable.
View return policy
Description
SNAP23 Polyclonal specifically detects SNAP23 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
SNAP23 | |
Polyclonal | |
Unconjugated | |
O00161 | |
SNAP23 | |
E. coli-derived recombinant human SNAP23 Asp146-Ser211 Accession # O00161 | |
100 μg | |
Primary | |
Detects human SNAP23 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse SNAP23 is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
HsT17016, SNAP-23, SNAP23A, SNAP23B, synaptosomal-associated protein 23, synaptosomal-associated protein, 23kD, synaptosomal-associated protein, 23kDa, Vesicle-membrane fusion protein SNAP-23 | |
Sheep | |
Affinity Purified | |
RUO | |
8773 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction