Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SNAP25 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$208.00 - $474.00
Specifications
Antigen | SNAP25 |
---|---|
Clone | 1065704 |
Dilution | Immunocytochemistry 8-25 μg/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11418100
![]() |
R&D Systems
MAB11418100 |
100 μg |
Each for $474.00
|
|
|||||
MAB11418SP
![]() |
R&D Systems
MAB11418SP |
25 μg |
Each for $208.00
|
|
|||||
Description
SNAP25 Monoclonal antibody specifically detects SNAP25 in Human samples. It is validated for ImmunocytochemistrySpecifications
SNAP25 | |
Immunocytochemistry 8-25 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
bA416N4.2, dJ1068F16.2, FLJ23079, resistance to inhibitors of cholinesterase 4 homolog, RIC4, RIC-4, SEC9, SNAP-25SNAP, SUP, Super protein, Synaptosomal-associated 25 kDa protein, synaptosomal-associated protein 25, synaptosomal-associated protein, 25kD, synaptosomal-associated protein, 25kDa | |
E. coli -derived recombinant human SNAP-25, Asp140-Gly206, Accession # P60880 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1065704 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P60880 | |
6616 | |
IgG3 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title