Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SNAP29 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7869SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7869SP 25 μg
MAB7869 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB7869SP Supplier R&D Systems Supplier No. MAB7869SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SNAP29 Monoclonal specifically detects SNAP29 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen SNAP29
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 827003
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunocytochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O95721
Gene Alias CEDNIK, CEDNIKSoluble 29 kDa NSF attachment protein, SNAP-29FLJ21051, synaptosomal-associated protein 29, synaptosomal-associated protein, 29kD, synaptosomal-associated protein, 29kDa, Vesicle-membrane fusion protein SNAP-29
Gene Symbols SNAP29
Host Species Mouse
Immunogen E. coli-derived recombinant human SNAP29 Met1-Glu129 Accession # O95721
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9342
Test Specificity Detects human SNAP29 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.