Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SNAPAP Partial ORF (NP_036569.1, 41 a.a. - 136 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7099585
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

SNAPAP is a component of the SNARE complex of proteins that is required for synaptic vesicle docking and fusion (Ilardi et al., 1999 [PubMed 10195194]). SNAPAP is also a component of the ubiquitously expressed BLOC1 multisubunit protein complex. BLOC1 is required for normal biogenesis of specialized organelles of the endosomal-lysosomal system, such as melanosomes and platelet dense granules (Starcevic and Dell'Angelica, 2004 [PubMed 15102850]).[supplied by OMIM]

Sequence: DSHVHAVRESQVELREQIDNLATELCRINEDQKVALDLDPYVKKLLNARRRVVLVNNILQNAQERLRRLNHSVAKETARRRAMLDSGIYPPGSPGK

Specifications

Accession Number NP_036569.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23557
Molecular Weight (g/mol) 36.3kDa
Name SNAPAP (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen DSHVHAVRESQVELREQIDNLATELCRINEDQKVALDLDPYVKKLLNARRRVVLVNNILQNAQERLRRLNHSVAKETARRRAMLDSGIYPPGSPGK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias SNAPAP
Common Name SNAPIN
Gene Symbol SNAPIN
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.