Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SNF1LK2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5737SP
This item is not returnable.
View return policy
Description
SNF1LK2/SIK2 Polyclonal specifically detects SNF1LK2/SIK2 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
SNF1LK2/SIK2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp434K1115, EC 2.7.11, EC 2.7.11.1, KIAA0781SIK-2, LOH11CR1I, QIK, QIKSNF1LK2, Qin-induced kinase, salt-inducible kinase 2, Salt-inducible protein kinase 2, salt-inducible serine/threonine kinase 2, serine/threonine-protein kinase SIK2, Serine/threonine-protein kinase SNF1-like kinase 2, SIK2, SNF1-like kinase 2 | |
Goat | |
Affinity Purified | |
RUO | |
23235 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Q9H0K1 | |
SIK2 | |
E. coli-derived recombinant human SNF1LK2 Ala663-Leu818 Accession # Q9H0K1 | |
25 μg | |
Primary | |
Detects human SNF1LK2 in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction