Learn More
Description
- Components:
- Biotinylated Detection Ab Diluent
- Concentrated Biotinylated Detection Ab (100×)
- Certificate of Analysis
- Concentrated HRP Conjugate (100×)
- HRP Conjugate Diluent
- Concentrated Wash Buffer (25×)
- Plate Sealer
- Micro ELISA Plate (Dismountable)
- Stop Solution
- Reference Standard
- Product Description
- Reference Standard & Sample Diluent
- Substrate Reagent
Specifications
Specifications
| Assay Sensitivity | 37.50 pg/mL |
| Product Type | ELISA |
| Specificity | This ELISA kit applies to the in vitro quantitative determination of Human SOD1 concentrations in serum, plasma and other biological fluids |
| Gene ID (Entrez) | 6647 |
| Gene Symbol | SOD1 |
| Host Species | Human |
| Quantity | 1 Kit |
| Sample Volume | 50 μL |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Target | SOD1/Cu-Zn SOD |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
