Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human SODD/BAG4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4350
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4350 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4350 Supplier R&D Systems Supplier No. AF4350
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

SODD/BAG4 Polyclonal specifically detects SODD/BAG4 in Human samples. It is validated for Western Blot, Immunocytochemistry, Knockout Validated.

Specifications

Antigen SODD/BAG4
Applications Western Blot, Immunocytochemistry, Immunoassay
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 μg/mL, Immunocytochemistry 5-15 μg/mL, Knockout Validated
Gene Accession No. O95429
Gene Alias BAG4, BAG-4, BAG-family molecular chaperone regulator-4, Bcl-2-associated athanogene 4, BCL2-associated athanogene 4, Silencer of death domains, SODDBAG family molecular chaperone regulator 4
Gene Symbols BAG4
Host Species Goat
Immunogen E. coli-derived recombinant human SODD/BAG4 Glu122-Leu457 Accession # O95429
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9530
Test Specificity Detects human SODD/BAG4 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.