Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Soggy-1/DkkL1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | Soggy-1/DkkL1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Soggy-1/DkkL1 Polyclonal specifically detects Soggy-1/DkkL1 in Human samples. It is validated for Western Blot.Specifications
Soggy-1/DkkL1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
Cancer/testis antigen 34, CT34dickkopf-like 1 (soggy), dickkopf-like 1, DkkL1, Protein soggy-1, SGY, SGY1, SGY-1dickkopf-like protein 1, SGY1soggy, soggy-1, Soggy1 | |
DKKL1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9UK85 | |
27120 | |
Mouse myeloma cell line NS0-derived recombinant human Soggy-1/DkkL1 Ala31-Leu242 Accession # Q9UK85 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Soggy-1/DkkL1 in direct ELISAs and Western blots. In Western blots, approximately 5% cross-reactivity with recombinant mouse Soggy-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title