Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Human Soluble Protein Master Buffer Kit

Catalog No. p-4003569
Click to view available options
No. of Tests:
100
500
Includes:
1 Bottle 30mL assay diluent, 1 bottle 5mL capture bead diluent, 1 bottle 5mL detection reagent diluent, 1 bottle 5mL capture bead diluent for Serum/Plasma, 1 bottle 130mL wash Buffer, 1 vial 0.25 mL instrument setup bead A1, 1 vial 0.25mL instrument setup bead A9, 1 vial 1.0mL instrument setup bead F1, 1 vial 0.25mL instrument setup bead F9, 1 vial, 0.25mL PE instrument setup bead F1, 1 vial 0.5mL PE positive control detector
1 bottle 150mL assay diluent, 1 bottle 30mL capture bead diluent, 1 bottle 30mL detection reagent diluent, 1 bottle 30mL capture bead diluent for Serum/Plasma, 1 bottle 650mL wash buffer, 1 vial 0.25 mL instrument setup bead A1, 1 vial 0.25mL instrument

Includes

  • Assay Diluent
  • Capture Bead Diluent
  • Detection Reagent Diluent
  • Capture Bead Diluent for Serum/Plasma
  • Wash Buffer
  • Instrument Setup Bead A1
  • Instrument Setup Bead A9
  • Instrument Setup Bead F1
  • Instrument Setup Bead F9
  • PE Instrument Setup Bead F1
  • PE Positive Control Detector

Along with Human soluble protein flex set system buffer kit employs particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations

Buffers and instrument setup reagents provided in this kit have been optimized for analysis of analytes in tissue culture supernatants, plasma, and serum samples.

  • Provides sufficient reagents for the quantitative analysis of 100 samples and 10 instrument setup procedures or 500 samples and 10 instrument setup procedures
  • Has been optimized for use with the CBA Human Soluble Protein Flex Sets
  • Working assay range for most analytes in this system is 10 to 2,500pg/mL

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cytometric Bead Array (CBA) Assay
Validated Application RUO
No. of Tests 100
Includes 1 Bottle 30mL assay diluent, 1 bottle 5mL capture bead diluent, 1 bottle 5mL detection reagent diluent, 1 bottle 5mL capture bead diluent for Serum/Plasma, 1 bottle 130mL wash Buffer, 1 vial 0.25 mL instrument setup bead A1, 1 vial 0.25mL instrument setup bead A9, 1 vial 1.0mL instrument setup bead F1, 1 vial 0.25mL instrument setup bead F9, 1 vial, 0.25mL PE instrument setup bead F1, 1 vial 0.5mL PE positive control detector
Content And Storage 2°C to 8°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.