Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Somatostatin R2/SSTR2 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC4224R100UG
This item is not returnable.
View return policy
Description
Somatostatin R2/SSTR2 Monoclonal specifically detects Somatostatin R2/SSTR2 in Human samples. It is validated for Flow Cytometry.Specifications
Somatostatin R2/SSTR2 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P30874 | |
SSTR2 | |
NS0 mouse myeloma cell line transfected with Somatostatin R2/SSTR2 Met1-Ile369 Accession # P30874 | |
RUO | |
6752 | |
Human | |
IgG2a |
Flow Cytometry | |
402038 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Smstr2, somatostatin receptor 2, somatostatin receptor type 2, SomatostatinR2, SRIF-1, SS2R, SS-2-R, SS2-R, SST2, SSTR2 | |
Mouse | |
100 μg | |
Primary | |
Detects human Somatostatin R2/SSTR2. Stains SSTR2 transfectants but not irrelevant transfectants. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction